Assay Details
Target Gene Details
Entrez Gene ID: | 14172 |
Gene Name: | fibroblast growth factor 18 |
Gene Aliases: |
D130055P09Rik, FGF-18 |
Location: |
Chr.11:33116970-33147400 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fgf18 | NM_008005.2 | 5 | 985 | NP_032031.1 |
NR_102395.1 | 5 | 1032 | ||
AF075291.1 | 5 | 993 | AAC62239.1 | |
AF211187.1 | 5 | 858 | AAF22976.1 | |
AK078245.1 | 5 | 851 |
More Information
Set Membership: |
Intragenic Exonic |