Assay Details
Target Gene Details
Entrez Gene ID: | 245827 |
Gene Name: | FAT tumor suppressor homolog 2 (Drosophila) |
Gene Aliases: |
EMI2, Fath2, Gm523, mKIAA0811 |
Location: |
Chr.11:55250609-55336621 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fat2 | NM_001029988.2 | 23 | 12855 | NP_001025159.1 |
AK132523.1 | 6 | 2078 | BAE21217.1 | |
AK173031.1 | 15 | 4753 | BAD32309.1 |
More Information
Set Membership: |
Intragenic Exonic |