Assay Details
Target Gene Details
Entrez Gene ID: | 72469 |
Gene Name: | phospholipase C, delta 3 |
Gene Aliases: |
2610205J15Rik, mKIAA1964 |
Location: |
Chr.11:103069236-103101658 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plcd3 | NM_152813.3 | 15 | 2912 | NP_690026.2 |
AK011892.1 | 11 | 1843 | ||
AK046669.1 | 15 | 2878 | BAC32829.1 | |
AK173311.1 | 13 | 4870 | BAD32589.1 | |
BC031392.1 | 15 | 2856 | AAH31392.1 | |
BY589869.1 | 1 | 330 |
More Information
Set Membership: |
Intragenic Exonic |