Assay Details
Target Gene Details
Entrez Gene ID: | 14104 |
Gene Name: | fatty acid synthase |
Gene Aliases: |
A630082H08Rik, FAS |
Location: |
Chr.11:120805958-120824547 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fasn | NM_007988.3 | 22 | 4204 | NP_032014.3 |
AF127033.1 | 22 | 3721 | AAG02285.1 | |
AK147214.1 | 22 | 3900 | BAE27771.1 | |
AK147694.1 | 22 | 3967 | ||
AK171499.1 | 22 | 3836 | BAE42492.1 | |
BC046513.1 | 22 | 3758 | AAH46513.1 |