Assay Details
Target Gene Details
Entrez Gene ID: | 11764 |
Gene Name: | adaptor protein complex AP-1, beta 1 subunit |
Gene Aliases: |
Adtb1, b2b1660Clo |
Location: |
Chr.11:4947521-5042794 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ap1b1 | NM_001243043.1 | 23 | 3740 | NP_001229972.1 |
NM_001243044.1 | 22 | 3522 | NP_001229973.1 | |
NM_007454.3 | 21 | 3513 | NP_031480.2 | |
AK034140.1 | 23 | 3743 | BAC28603.1 | |
AK155892.1 | 21 | 3433 | BAE33487.1 | |
AK159273.1 | 21 | 3404 | BAE34952.1 | |
AK160042.1 | 21 | 3404 | BAE35584.1 | |
AK165747.1 | 22 | 3441 | ||
AK170376.1 | 21 | 3424 | ||
AK213130.1 | 1 | 266 | ||
BC008513.1 | 21 | 3326 | AAH08513.1 | |
BC066827.1 | 12 | 2057 | AAH66827.1 | |
BP765289.1 | 1 | 597 | ||
Y07919.1 | 21 | 3368 | CAA69224.1 |
More Information
Set Membership: |
Intragenic Exonic |