Assay Details
Target Gene Details
Entrez Gene ID: | 73635 |
Gene Name: | prostaglandin E synthase 3 (cytosolic)-like |
Gene Aliases: |
1110069E20Rik, 1700113I22Rik |
Location: |
Chr.11:101418801-101425333 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptges3l | NM_026865.2 | 6 | 1218 | NP_081141.1 |
AK007198.1 | 6 | 1217 | BAB24896.1 |
More Information
Set Membership: |
Intragenic Exonic |