Assay Details
Target Gene Details
Entrez Gene ID: | 208990 |
Gene Name: | neuropeptide B |
Gene Aliases: |
- |
Location: |
Chr.11:120608477-120609100 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npb | NM_153288.3 | 1 | 131 | NP_695020.1 |
AB085945.1 | 1 | 99 | BAC07178.1 | |
AK141698.1 | 1 | 132 | BAE24805.1 | |
BC134361.1 | 1 | 103 |