Assay Details
Target Gene Details
Entrez Gene ID: | 72055 |
Gene Name: | solute carrier family 38, member 10 |
Gene Aliases: |
1810073N04Rik |
Location: |
Chr.11:120103951-120151351 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc38a10 | NM_001164798.1 | 17 | 2848 | NP_001158270.1 |
NM_001164799.1 | 16 | 2827 | NP_001158271.1 | |
NM_001164800.1 | 16 | 2824 | NP_001158272.1 | |
NM_024249.5 | 17 | 2851 | NP_077211.4 | |
AK032245.1 | 17 | 2775 | BAC27777.1 | |
AK159948.1 | 16 | 2789 | BAE35504.1 | |
AK165672.1 | 16 | 2791 | ||
AK215358.1 | 1 | 212 | ||
BC005641.1 | 5 | 1200 | AAH05641.1 | |
BC023375.1 | AAH23375.1 | |||
BC043299.1 | 7 | 1524 | AAH43299.1 | |
BC055017.1 | 4 | 872 | AAH55017.1 | |
BC082300.1 | 16 | 2795 | AAH82300.1 | |
BC088811.1 | 17 | 2784 | AAH88811.1 |
More Information
Set Membership: |
Intragenic Exonic |