Assay Details
Target Gene Details
Entrez Gene ID: | 268390 |
Gene Name: | AHA1, activator of heat shock protein ATPase 2 |
Gene Aliases: |
- |
Location: |
Chr.11:23487882-23498039 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ahsa2 | NM_001290654.1 | 9 | 2344 | NP_001277583.1 |
NM_001290655.1 | 9 | 2368 | NP_001277584.1 | |
NM_172391.4 | 9 | 2349 | NP_765979.3 | |
AK035941.1 | 9 | 2122 | BAC29251.1 | |
AK093945.1 | 9 | 2256 | BAC04256.1 | |
AK137116.1 | 9 | 2260 | BAE23240.1 | |
BB103167.2 | ||||
BC038397.1 | 9 | 2235 | ||
BC064012.1 | 9 | 2169 | AAH64012.1 |
Target Gene Details
Entrez Gene ID: | 17847 |
Gene Name: | ubiquitin specific peptidase 34 |
Gene Aliases: |
A530081C03Rik, Murr2 |
Location: |
Chr.11:23306846-23490560 on Build GRCm38 |
Assay Gene Location: | Within Exon 80 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Usp34 | NM_001190401.2 | 80 | 10870 | NP_001177330.2 |
AK129165.1 | 26 | 3709 | BAC97975.1 |
More Information
Set Membership: |
Intragenic Exonic |