Assay Details
Target Gene Details
Entrez Gene ID: | 18604 |
Gene Name: | pyruvate dehydrogenase kinase, isoenzyme 2 |
Gene Aliases: |
- |
Location: |
Chr.11:95026258-95041371 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdk2 | NM_133667.2 | 11 | 2088 | NP_598428.2 |
AF267660.1 | 11 | 2049 | AAF72038.1 | |
AI848783.1 | 1 | 192 | ||
AK002647.1 | 11 | 2032 | ||
BC021764.1 | 11 | 2042 | AAH21764.1 | |
CX204954.1 | 1 | 592 |
More Information
Set Membership: |
Intragenic Exonic |