Assay Details
Target Gene Details
Entrez Gene ID: | 171285 |
Gene Name: | hepatitis A virus cellular receptor 2 |
Gene Aliases: |
TIM-3, Tim3, Timd3 |
Location: |
Chr.11:46454547-46481255 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Havcr2 | NM_134250.2 | 7 | 1087 | NP_599011.2 |
AF450241.1 | 7 | 1083 | AAL65156.1 | |
AK148474.1 | 7 | 1088 | BAE28574.1 | |
AK170775.1 | 8 | 908 | ||
BC106851.2 | 7 | 1045 | AAI06852.1 | |
BC106852.2 | 7 | 1045 | AAI06853.1 |
More Information
Set Membership: |
Intragenic Exonic |