Assay Details
Target Gene Details
Entrez Gene ID: | 56218 |
Gene Name: | POZ (BTB) and AT hook containing zinc finger 1 |
Gene Aliases: |
8430401L15Rik, AI447519, Mazr, Patz, Zfp278 |
Location: |
Chr.11:3289131-3309083 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Patz1 | NM_001253690.1 | 5 | 1883 | NP_001240619.1 |
NM_001253691.1 | 5 | 1902 | NP_001240620.1 | |
NM_019574.3 | 4 | 1810 | NP_062520.1 | |
NR_045108.2 | 6 | 764 | ||
NR_045109.2 | 4 | 599 | ||
AB029397.1 | 4 | 1810 | BAA90874.1 | |
AK018372.1 | 4 | 564 | ||
BC026673.1 | 4 | 481 | AAH26673.1 | |
BC043035.1 | 3 | 1636 | AAH43035.1 | |
BC055947.1 | 6 | 614 | AAH55947.1 | |
CA327687.1 | 5 | 491 |
More Information
Set Membership: |
Intragenic Exonic |