Assay Details
Target Gene Details
Entrez Gene ID: | 11764 |
Gene Name: | adaptor protein complex AP-1, beta 1 subunit |
Gene Aliases: |
Adtb1, b2b1660Clo |
Location: |
Chr.11:4947521-5042794 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ap1b1 | NM_001243043.1 | 1 | 369 | NP_001229972.1 |
AK034140.1 | 1 | 370 | BAC28603.1 |
Target Gene Details
Entrez Gene ID: | 380684 |
Gene Name: | neurofilament, heavy polypeptide |
Gene Aliases: |
NF-H, NF200, Nfh, mKIAA0845 |
Location: |
Chr.11:4938754-4948064 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nefh | NM_010904.3 | 1 | 176 | NP_035034.2 |
AK122388.1 | 1 | 78 | BAC65670.1 | |
M35131.1 | 2 | 200 | AAA39809.1 |
More Information
Set Membership: |
Intragenic Exonic |