Assay Details
Target Gene Details
Entrez Gene ID: | 320563 |
Gene Name: | immunoglobulin superfamily containing leucine-rich repeat 2 |
Gene Aliases: |
B930052A04Rik, Linx, Mbu-3, mKIAA1465 |
Location: |
Chr.9:58196297-58204319 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Islr2 | NM_001161535.1 | 2 | 2897 | NP_001155007.1 |
NM_001161536.1 | 4 | 3078 | NP_001155008.1 | |
NM_001161537.1 | 3 | 3069 | NP_001155009.1 | |
NM_001161538.1 | 4 | 2846 | NP_001155010.1 | |
NM_001161539.1 | 4 | 2808 | NP_001155011.1 | |
NM_001161540.1 | 3 | 2764 | NP_001155012.1 | |
NM_001161541.1 | 3 | 2655 | NP_001155013.1 | |
NM_177193.5 | 3 | 2888 | NP_796167.2 | |
AK043849.1 | 1 | 722 | ||
AK047348.1 | 1 | 2376 | BAC33030.1 | |
AK129367.1 | 3 | 2574 | BAC98177.1 | |
BC059068.1 | 3 | 2741 | AAH59068.1 | |
BC096531.1 | 2 | 2897 | AAH96531.1 | |
EU497918.1 | 3 | 2705 |
More Information
Set Membership: |
Intragenic Exonic |