Assay Details
Target Gene Details
Entrez Gene ID: | 74840 |
Gene Name: | mesencephalic astrocyte-derived neurotrophic factor |
Gene Aliases: |
3230402M22Rik, AA407711, AA408789, AA673178, Armet, D17914, D18Mgi17 |
Location: |
Chr.9:106868310-106891985 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Manf |
Target Gene Details
Entrez Gene ID: | 321006 |
Gene Name: | Vpr (HIV-1) binding protein |
Gene Aliases: |
AI447437, B930007L02Rik, mKIAA0800 |
Location: |
Chr.9:106814172-106880992 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Vprbp | NM_001015507.2 | 23 | 4500 | NP_001015507.1 |
AK029372.1 | 7 | 845 | BAC26425.1 | |
AK041367.1 | 1 | 518 | ||
AK080996.1 | 1 | 2186 | ||
AK135721.1 | BAE22628.1 | |||
AK164773.1 | 1 | 1595 | ||
BC065119.1 | 23 | 4500 | AAH65119.1 |