Assay Details
Target Gene Details
Entrez Gene ID: | 56533 |
Gene Name: | regulator of G-protein signaling 17 |
Gene Aliases: |
6430507P11Rik, Rgsz2 |
Location: |
Chr.10:5825663-5922400 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rgs17 | NM_001161822.1 | 6 | 5698 | NP_001155294.1 |
NM_019958.4 | 5 | 5628 | NP_064342.1 |
More Information
Set Membership: |
Intragenic Exonic |