Assay Details
Target Gene Details
Entrez Gene ID: | 237320 |
Gene Name: | aldehyde dehydrogenase 8 family, member A1 |
Gene Aliases: |
Raldh4 |
Location: |
Chr.10:21377300-21396578 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aldh8a1 | NM_178713.4 | 7 | 1449 | NP_848828.1 |
AF510322.1 | 7 | 1458 | AAO64246.1 | |
AK050298.1 | 7 | 1450 | BAC34173.1 | |
AK143752.1 | 7 | 1450 | BAE25524.1 | |
BC013511.1 | 5 | 991 | AAH13511.1 | |
BC038493.1 | 7 | 1461 | AAH38493.1 |
More Information
Set Membership: |
Intragenic Exonic |