Assay Details
Target Gene Details
Entrez Gene ID: | 19714 |
Gene Name: | REV3-like, catalytic subunit of DNA polymerase zeta RAD54 like (S. cerevisiae) |
Gene Aliases: |
Rev, Rev3, Sez4 |
Location: |
Chr.10:39732099-39875210 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rev3l | NM_011264.3 | 13 | 2608 | NP_035394.2 |
AB031049.1 | 13 | 2609 | BAA90768.1 | |
AF083464.1 | 14 | 2341 | AAC98785.1 | |
AK032532.1 | 13 | 2652 | ||
AK164702.1 | BAE37883.1 |
More Information
Set Membership: |
Intragenic Exonic |