Assay Details
Target Gene Details
Entrez Gene ID: | 102634101 |
Gene Name: | predicted gene, 31763 |
Gene Aliases: |
- |
Location: |
Chr.10:67005075-67041326 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm31763 | NR_110511.1 | |||
AK079018.1 |
Target Gene Details
Entrez Gene ID: | 28193 |
Gene Name: | receptor accessory protein 3 |
Gene Aliases: |
D10Ucla1 |
Location: |
Chr.10:67009189-67096988 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Reep3 | NM_001204915.1 | 9 | 1335 | NP_001191844.1 |
NM_178606.5 | 8 | 1258 | NP_848721.1 | |
AK005026.1 | 8 | 1204 | ||
AK012403.1 | 8 | 1202 | BAB28218.1 | |
AK028868.1 | 8 | 1215 | ||
AK047728.1 | 8 | 1126 | BAC33141.1 | |
AK079649.1 | 8 | 1128 | BAC37714.1 | |
AK154559.1 | 9 | 1193 | ||
BC004607.1 | 8 | 1167 | AAH04607.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |