Assay Details
Target Gene Details
Entrez Gene ID: | 118445 |
Gene Name: | Kruppel-like factor 16 |
Gene Aliases: |
AI843742, BTEB4, DRRF |
Location: |
Chr.10:80567121-80577296 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klf16 | NM_078477.2 | 1 | 376 | NP_510962.2 |
AF283891.1 | 1 | 374 | AAK66968.1 | |
AK044577.1 | 1 | 377 | BAC31987.1 | |
AK154524.1 | 1 | 403 | BAE32651.1 | |
BC137607.1 | 1 | 309 | ||
BC137609.1 | 1 | 309 |
More Information
Set Membership: |
Intragenic Exonic |