Assay Details
Target Gene Details
Entrez Gene ID: | 104248 |
Gene Name: | calcineurin binding protein 1 |
Gene Aliases: |
A330070M20Rik, Cain, Ppp3in |
Location: |
Chr.10:75646110-75764358 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cabin1 | NM_172549.3 | 23 | 3897 | NP_766137.2 |
AK078480.1 | BAC37299.1 | |||
BC057551.1 | AAH57551.1 | |||
BC150825.1 | 22 | 3461 | ||
BU705499.1 | 3 | 423 |
More Information
Set Membership: |
Intragenic Exonic |