Assay Details
Target Gene Details
Entrez Gene ID: | 103213 |
Gene Name: | TRAF3 interacting protein 2 |
Gene Aliases: |
AI429613, Act1, CIKS |
Location: |
Chr.10:39612934-39655307 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Traf3ip2 | NM_134000.3 | 1 | 136 | NP_598761.2 |
AK037407.1 | BAC29801.1 | |||
AK052190.1 | 1 | 136 | BAC34875.1 | |
JX869941.1 | 1 | 143 |
More Information
Set Membership: |
Intragenic Exonic |