Assay Details
Target Gene Details
Entrez Gene ID: | 57014 |
Gene Name: | 5-hydroxytryptamine (serotonin) receptor 3B |
Gene Aliases: |
5-HT3B |
Location: |
Chr.9:48935008-48964990 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Htr3b | NM_020274.4 | 9 | 1480 | NP_064670.1 |
AF155045.1 | 9 | 1312 | AAF73284.1 | |
AK085213.1 | 9 | 1481 | BAC39391.1 | |
BC046990.1 | 9 | 1458 | AAH46990.1 |
More Information
Set Membership: |
Intragenic Exonic |