Assay Details
Target Gene Details
Entrez Gene ID: | 16000 |
Gene Name: | insulin-like growth factor 1 |
Gene Aliases: |
C730016P09Rik, Igf-1, Igf-I |
Location: |
Chr.10:87858270-87937047 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Igf1 | NM_001111275.1 | 1 | 117 | NP_001104745.1 |
NM_010512.4 | 1 | 117 | NP_034642.2 | |
AK050118.1 | 1 | 106 | ||
AK081019.1 | 1 | 106 | BAC38117.1 | |
AK155435.1 | BAE33264.1 | |||
AK165471.1 | BAE38208.1 |
More Information
Set Membership: |
Intragenic Exonic |