Assay Details
Target Gene Details
Entrez Gene ID: | 74048 |
Gene Name: | RIKEN cDNA 4632428N05 gene |
Gene Aliases: |
Dies1, PD-1H, VISTA |
Location: |
Chr.10:60346851-60428381 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4632428N05Rik | NM_001159572.1 | 7 | 2948 | NP_001153044.1 |
NM_028732.4 | 7 | 2951 | NP_083008.1 | |
AK014600.1 | 7 | 2879 | BAB29455.1 | |
AK032383.1 | 7 | 2894 | BAC27847.1 | |
AK033393.1 | 7 | 2897 | ||
AK079734.1 | 7 | 2899 | BAC37735.1 | |
AK131118.1 | 7 | 2864 | BAD21368.1 | |
AK142494.1 | 1 | 797 | ||
AK163118.1 | 7 | 2896 | BAE37201.1 | |
AK170419.1 | 7 | 2896 | BAE41785.1 | |
AK171045.1 | 3 | 994 |
Target Gene Details
Entrez Gene ID: | 22295 |
Gene Name: | cadherin 23 (otocadherin) |
Gene Aliases: |
4930542A03Rik, USH1D, ahl, ahl1, bob, bus, mdfw, nmf112, nmf181, nmf252, sals, v |
Location: |
Chr.10:60302414-60696515 on Build GRCm38 |
Assay Gene Location: | Within Intron 39 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdh23 | NM_001252635.1 | NP_001239564.1 | ||
NM_023370.3 | NP_075859.2 | |||
NR_045556.1 | ||||
AF308939.1 | AAG52817.1 | |||
AJ888000.1 | CAI59609.1 | |||
AJ888001.1 | CAI59610.1 | |||
AJ888002.1 | CAI59611.1 | |||
AY026062.1 | AAK07670.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |