Assay Details
Target Gene Details
Entrez Gene ID: | 74840 |
Gene Name: | mesencephalic astrocyte-derived neurotrophic factor |
Gene Aliases: |
3230402M22Rik, AA407711, AA408789, AA673178, Armet, D17914, D18Mgi17 |
Location: |
Chr.9:106868310-106891985 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Manf | NM_029103.3 | 4 | 1912 | NP_083379.2 |
AK131997.1 | 4 | 1913 | BAE20927.1 |