Assay Details
Target Gene Details
Entrez Gene ID: | 67569 |
Gene Name: | MGAT4 family, member C |
Gene Aliases: |
9130411I17Rik, GntIVh |
Location: |
Chr.10:101681487-102391851 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mgat4c | NM_001162368.1 | 6 | 1160 | NP_001155840.1 |
NM_001162369.1 | 6 | 1033 | NP_001155841.1 | |
NM_001205098.1 | 8 | 1573 | NP_001192027.1 | |
NM_026243.4 | 4 | 1055 | NP_080519.2 | |
AK018574.1 | 4 | 1054 | BAB31286.1 | |
AK018672.1 | 4 | 1058 | BAB31336.1 | |
AK032009.1 | 6 | 1034 | ||
AK033482.1 | 4 | 1056 | BAC28312.1 | |
AK033643.1 | 1 | 310 | ||
BC046987.1 | 4 | 1043 | AAH46987.1 |
More Information
Set Membership: |
Intragenic Exonic |