Assay Details
Target Gene Details
Entrez Gene ID: | 15353 |
Gene Name: | high mobility group 20B |
Gene Aliases: |
AW610687, BRAF35, Hmgx2, Hmgxb2, Smarce1r |
Location: |
Chr.10:81346046-81350511 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hmg20b | NM_001163165.1 | 1 | 100 | NP_001156637.1 |
NM_001163166.1 | NP_001156638.1 | |||
NM_010440.3 | 1 | 101 | NP_034570.1 | |
AF146224.1 | 1 | 84 | AAF66708.1 | |
AK153816.1 | BAE32194.1 | |||
AK155615.1 | 1 | 101 | BAE33346.1 | |
AK155652.1 | BAE33368.1 | |||
AK155890.1 | BAE33486.1 | |||
AL355734.1 | 1 | 96 | CAB90813.1 | |
BC011334.1 | AAH11334.1 |