Assay Details
Target Gene Details
Entrez Gene ID: | 19263 |
Gene Name: | protein tyrosine phosphatase, receptor type, B |
Gene Aliases: |
3230402H02Rik, C130094E24, Ptpz, Rptpb, VE-PTP, Veptp |
Location: |
Chr.10:116275516-116389538 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprb | NM_029928.2 | 17 | 4366 | NP_084204.2 |
AK147439.1 | 17 | 4368 | BAE27912.1 | |
AK147668.1 | 17 | 4368 | BAE28060.1 | |
AY077755.1 | 17 | 4244 | AAL75813.1 | |
BC047086.1 | 7 | 1426 | AAH47086.1 | |
BC141006.1 | 17 | 4162 | ||
BC145111.1 | 17 | 4162 |
More Information
Set Membership: |
Intragenic Exonic |