Assay Details
Target Gene Details
Entrez Gene ID: | 117599 |
Gene Name: | helicase (DNA) B |
Gene Aliases: |
AI447783, D10Ertd664e |
Location: |
Chr.10:120083608-120112965 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Helb | NM_080446.2 | 12 | 3004 | NP_536694.2 |
AB048542.1 | 12 | 2929 | BAB20809.1 | |
AK131122.1 | 12 | 3026 | BAD21372.1 | |
BC068140.1 | 12 | 2932 | AAH68140.1 |
More Information
Set Membership: |
Intragenic Exonic |