Assay Details
Target Gene Details
Entrez Gene ID: | 23988 |
Gene Name: | protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1 |
Gene Aliases: |
0610025L01Rik, D9Bwg1161e |
Location: |
Chr.9:20652130-20666584 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pin1 | NM_023371.3 | 4 | 3226 | NP_075860.1 |
AK054045.1 | 4 | 3227 | BAC35631.1 |
More Information
Set Membership: |
Intragenic Exonic |