Assay Details
Target Gene Details
Entrez Gene ID: | 171167 |
Gene Name: | fucosyltransferase 10 |
Gene Aliases: |
fuc-TX, fut10A |
Location: |
Chr.8:31187331-31261924 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fut10 | NM_001012517.5 | 3 | 520 | NP_001012535.1 |
NM_001286422.1 | 4 | 625 | NP_001273351.1 | |
NM_001286424.1 | 3 | 413 | NP_001273353.1 | |
NM_001286425.1 | 3 | 520 | NP_001273354.1 | |
NM_134161.3 | 3 | 520 | NP_598922.1 | |
AJ880009.1 | 2 | 133 | CAI53944.1 | |
AK083041.1 | 3 | 509 | BAC38741.1 | |
AK147136.1 | 4 | 581 | BAE27706.1 | |
AK169153.1 | 3 | 474 | BAE40932.1 | |
AK178457.1 | ||||
BC030474.1 | 3 | 417 | AAH30474.1 | |
BC062113.1 | 3 | 519 | AAH62113.1 | |
BY721538.1 | 3 | 521 |
More Information
Set Membership: |
Intragenic Exonic |