Assay Details
Target Gene Details
Entrez Gene ID: | 114255 |
Gene Name: | docking protein 4 |
Gene Aliases: |
- |
Location: |
Chr.8:94863828-94876312 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dok4 | NM_053246.3 | 9 | 2373 | NP_444476.1 |
AK077612.1 | 9 | 2365 | ||
AK139363.1 | 8 | 2262 | ||
AK142621.1 | 8 | 2280 | BAE25134.1 | |
AK156485.1 | 9 | 2373 | BAE33728.1 | |
BC004705.1 | 9 | 2334 | AAH04705.1 |
Target Gene Details
Entrez Gene ID: | 20021 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide C |
Gene Aliases: |
33kDa, RPB3, Rpo2-3, mRBP31 |
Location: |
Chr.8:94857450-94864242 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Polr2c | NM_009090.5 | 9 | 1397 | NP_033116.2 |
BC099548.1 | 9 | 1342 | AAH99548.1 |
More Information
Set Membership: |
Intragenic Exonic |