Assay Details
Target Gene Details
Entrez Gene ID: | 102247 |
Gene Name: | 1-acylglycerol-3-phosphate O-acyltransferase 6 (lysophosphatidic acid acyltransferase, zeta) |
Gene Aliases: |
AU041707, AW545732, GPAT4, Tsarg7 |
Location: |
Chr.8:23171265-23209171 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Agpat6 | NM_018743.4 | 13 | 3377 | NP_061213.2 |
AK045235.1 | 13 | 3271 | BAC32273.1 | |
AK083589.1 | 13 | 3271 | BAC38962.1 |
More Information
Set Membership: |
Intragenic Exonic |