Assay Details
Target Gene Details
Entrez Gene ID: | 68969 |
Gene Name: | eukaryotic translation initiation factor 1B |
Gene Aliases: |
1500010M16Rik, 3110001N14Rik, Gc20 |
Location: |
Chr.9:120492606-120495327 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif1b | NM_026892.3 | 4 | 634 | NP_081168.1 |
AK005192.1 | 4 | 601 | BAB23874.1 | |
AK013965.1 | 4 | 607 | BAB29089.1 | |
BC030319.1 | 4 | 503 | AAH30319.1 | |
BC033505.1 | 4 | 605 | AAH33505.1 |
More Information
Set Membership: |
Intragenic Exonic |