Assay Details
Target Gene Details
Entrez Gene ID: | 14182 |
Gene Name: | fibroblast growth factor receptor 1 |
Gene Aliases: |
AW208770, Eask, FLG, Fgfr-1, Flt-2, Hspy, MFR, bFGF-R-1, c-fgr |
Location: |
Chr.8:25513623-25575718 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fgfr1 | NM_001079908.2 | 1 | 338 | NP_001073377.1 |
NM_001079909.2 | 1 | 338 | NP_001073378.1 | |
NM_010206.3 | 1 | 338 | NP_034336.2 | |
AK049704.1 | 1 | 306 | ||
BC010200.1 | 1 | 237 | AAH10200.1 | |
BU611199.1 | 1 | 325 | ||
CB524826.1 | 1 | 325 | ||
CF532036.1 | 1 | 338 | ||
S74765.1 | 1 | 302 | AAB32844.2 |
More Information
Set Membership: |
Intragenic Exonic |