Assay Details
Target Gene Details
Entrez Gene ID: | 73711 |
Gene Name: | multivesicular body subunit 12A |
Gene Aliases: |
1110012M11Rik, Fam125a |
Location: |
Chr.8:71542880-71548026 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mvb12a | NM_028617.2 | 9 | 1018 | NP_082893.1 |
AK003644.1 | 9 | 998 | ||
BC020119.1 | 9 | 1018 | AAH20119.1 | |
BC027419.1 | 9 | 994 | AAH27419.1 | |
BC087734.1 | 9 | 1015 | AAH87734.1 |