Assay Details
Target Gene Details
Entrez Gene ID: | 407789 |
Gene Name: | cDNA sequence BC048644 |
Gene Aliases: |
- |
Location: |
Chr.8:121907833-121918428 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BC048644 | NM_001033485.2 | NP_001028657.1 | ||
AK133128.1 | BAE21521.1 |
Target Gene Details
Entrez Gene ID: | 12352 |
Gene Name: | carbonic anhydrase 5a, mitochondrial |
Gene Aliases: |
CAV, Ca5a, Car5 |
Location: |
Chr.8:121916135-121944912 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Car5a | NM_007608.2 | 7 | 1008 | NP_031634.2 |
AI132225.1 | 1 | 241 | ||
AI527458.1 | 1 | 229 | ||
BC030174.1 | 7 | 986 | AAH30174.1 | |
X51971.1 | 7 | 997 | CAA36233.1 |