Assay Details
Target Gene Details
Entrez Gene ID: | 235599 |
Gene Name: | RIKEN cDNA 6430571L13 gene |
Gene Aliases: |
- |
Location: |
Chr.9:107340640-107349683 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
6430571L13Rik | NM_175486.3 | 3 | 570 | NP_780695.1 |
AK032358.1 | 3 | 571 | BAC27833.1 | |
AK032501.1 | 3 | 577 | BAC27899.1 | |
AK043338.1 | 3 | 610 | BAC31524.1 | |
AK085251.1 | 3 | 601 | BAC39401.1 | |
BC115775.1 | 3 | 482 | ||
BC116708.1 | 3 | 482 |