Assay Details
Target Gene Details
Entrez Gene ID: | 17962 |
Gene Name: | N-acetyltransferase 3 |
Gene Aliases: |
- |
Location: |
Chr.8:67523854-67548627 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nat3 | NM_008674.2 | 3 | 1010 | NP_032700.1 |
AK077364.1 | 3 | 1010 | BAC36769.1 |
More Information
Set Membership: |
Intragenic Exonic |