Assay Details
Target Gene Details
Entrez Gene ID: | 68119 |
Gene Name: | CKLF-like MARVEL transmembrane domain containing 3 |
Gene Aliases: |
9430096L06Rik, AI413895, BNAS2, Cklfsf3 |
Location: |
Chr.8:104339383-104347672 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cmtm3 | NM_024217.3 | 1 | 54 | NP_077179.1 |
AK079165.1 | BAC37565.1 | |||
AK172091.1 | BAE42821.1 | |||
AY241870.1 | 1 | 54 | AAP33492.1 | |
BC003230.1 | AAH03230.1 |
More Information
Set Membership: |
Intragenic Exonic |