Assay Details
Target Gene Details
Entrez Gene ID: | 26386 |
Gene Name: | heat shock transcription factor 4 |
Gene Aliases: |
HSF 4, HSTF 4, HSTF4, ldis1, mHSF4 |
Location: |
Chr.8:105269748-105275845 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hsf4 | NM_001256042.1 | 13 | 1686 | NP_001242971.1 |
NM_001256044.1 | 12 | 1577 | NP_001242973.1 | |
NM_011939.3 | 13 | 1596 | NP_036069.1 | |
NR_045689.1 | 13 | 1582 | ||
AB029349.1 | 13 | 1618 | BAA84583.1 | |
AB029350.1 | 13 | 1523 | BAA84584.1 | |
AK148056.1 | BAE28316.1 |
More Information
Set Membership: |
Intragenic Exonic |