Assay Details
Target Gene Details
Entrez Gene ID: | 12769 |
Gene Name: | chemokine (C-C motif) receptor 9 |
Gene Aliases: |
A130091K22Rik, Cmkbr10, GPR-9-6 |
Location: |
Chr.9:123767211-123783457 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccr9 | NM_001166625.1 | 4 | 1998 | NP_001160097.1 |
NM_009913.6 | 3 | 1930 | NP_034043.1 | |
AJ131357.2 | 4 | 1935 | CAB66136.2 | |
AK050615.1 | 3 | 1931 | BAC34344.1 | |
AK153975.1 | 3 | 1931 | BAE32291.1 | |
BC132166.1 | 2 | 1839 | ||
BC132466.1 | 2 | 1839 |