Assay Details
Target Gene Details
Entrez Gene ID: | 12776 |
Gene Name: | chemokine (C-C motif) receptor 8 |
Gene Aliases: |
C-C, C-C CKR-8, CC-CKR-8, CCR-8, CKR-8, Cmkbr8, mCCR8 |
Location: |
Chr.9:120092133-120094906 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccr8 | NM_007720.2 | 2 | 787 | NP_031746.1 |
BC101966.1 | 1 | 731 | AAI01967.1 | |
BC101967.1 | 1 | 731 | AAI01968.1 | |
BC103566.1 | 1 | 731 | AAI03567.1 | |
BC103567.1 | 1 | 731 | AAI03568.1 |