Assay Details
Target Gene Details
Entrez Gene ID: | 12768 |
Gene Name: | chemokine (C-C motif) receptor 1 |
Gene Aliases: |
Cmkbr1, Mip-1a-R |
Location: |
Chr.9:123962126-123968692 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccr1 | NM_009912.4 | 2 | 802 | NP_034042.3 |
AK031109.1 | 2 | 803 | BAC27256.1 | |
AK036597.1 | 2 | 504 | BAC29500.1 | |
AK036690.1 | 2 | 802 | BAC29535.1 | |
AK076275.1 | 2 | 677 | BAC36284.1 | |
BC011092.1 | 2 | 505 | AAH11092.1 | |
CT010178.1 | 1 | 337 | CAJ18386.1 | |
U29678.1 | 1 | 337 | AAA86119.1 |