Assay Details
Target Gene Details
Entrez Gene ID: | 102247 |
Gene Name: | 1-acylglycerol-3-phosphate O-acyltransferase 6 (lysophosphatidic acid acyltransferase, zeta) |
Gene Aliases: |
AU041707, AW545732, GPAT4, Tsarg7 |
Location: |
Chr.8:23171265-23209171 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Agpat6 | NM_018743.4 | 4 | 1255 | NP_061213.2 |
AF406611.1 | 3 | 766 | AAP97283.1 | |
AK045235.1 | 4 | 1149 | BAC32273.1 | |
AK049675.1 | 4 | 1149 | ||
AK077561.1 | 4 | 1139 | BAC36864.1 | |
AK083589.1 | 4 | 1149 | BAC38962.1 | |
AK161270.1 | 4 | 1118 | BAE36282.1 | |
AK162033.1 | 1 | 60 | BAE36691.1 | |
AK167876.1 | 4 | 1164 | BAE39890.1 | |
AK169257.1 | 4 | 1149 | BAE41020.1 | |
AK178773.1 | 3 | 244 | ||
AK182757.1 | 3 | 304 | ||
AK188390.1 | 3 | 363 | ||
AK204634.1 | 3 | 303 | ||
AY489184.1 | 3 | 324 | AAS75838.1 | |
BC031767.1 | 3 | 609 | AAH31767.1 |
More Information
Set Membership: |
Intragenic Exonic |