Assay Details
Target Gene Details
Entrez Gene ID: | 102247 |
Gene Name: | 1-acylglycerol-3-phosphate O-acyltransferase 6 (lysophosphatidic acid acyltransferase, zeta) |
Gene Aliases: |
AU041707, AW545732, GPAT4, Tsarg7 |
Location: |
Chr.8:23171265-23209171 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Agpat6 | NM_018743.4 | 2 | 969 | NP_061213.2 |
AF406611.1 | 1 | 481 | AAP97283.1 | |
AK045235.1 | 2 | 863 | BAC32273.1 | |
AK049675.1 | 2 | 863 | ||
AK077561.1 | 2 | 849 | BAC36864.1 | |
AK083589.1 | 2 | 863 | BAC38962.1 | |
AK161270.1 | 2 | 833 | BAE36282.1 | |
AK167876.1 | 2 | 878 | BAE39890.1 | |
AK169257.1 | 2 | 863 | BAE41020.1 | |
AK178773.1 | ||||
AK182757.1 | ||||
AK183848.1 | ||||
AK188390.1 | 1 | 76 | ||
AK192499.1 | ||||
AK204634.1 | ||||
AY489184.1 | AAS75838.1 | |||
BC031767.1 | 1 | 323 | AAH31767.1 |
More Information
Set Membership: |
Intragenic Exonic |