Assay Details
Target Gene Details
Entrez Gene ID: | 233876 |
Gene Name: | HIRA interacting protein 3 |
Gene Aliases: |
B130036O03, C86302 |
Location: |
Chr.7:126861972-126865377 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hirip3 | NM_172746.3 | 1 | 363 | NP_766334.1 |
BC055687.1 | 1 | 363 | AAH55687.1 |
Target Gene Details
Entrez Gene ID: | 233875 |
Gene Name: | INO80 complex subunit E |
Gene Aliases: |
AI225782, AI854876, Ccdc85, Ccdc95 |
Location: |
Chr.7:126851584-126862371 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ino80e | BC055928.1 | AAH55928.1 |