Assay Details
Target Gene Details
Entrez Gene ID: | 69920 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide I |
Gene Aliases: |
2810002B19Rik |
Location: |
Chr.7:30232074-30233387 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Polr2i | NM_027259.1 | 1 | 164 | NP_081535.1 |
BC062812.1 | 1 | 164 | AAH62812.1 |
More Information
Set Membership: |
Intragenic Exonic |