Assay Details
Target Gene Details
Entrez Gene ID: | 269952 |
Gene Name: | GDP-D-glucose phosphorylase 1 |
Gene Aliases: |
D330012F22Rik |
Location: |
Chr.7:80232893-80241420 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gdpgp1 | NM_178752.3 | 2 | 2439 | NP_848867.2 |
AK084531.1 | 2 | 2425 | BAC39213.1 | |
AK143704.1 | 2 | 2465 | BAE25508.1 |